Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676493

Sigma-Aldrich

VEGF Inhibitor, Flt211

The VEGF Inhibitor, Flt2-11 controls the biological activity of VEGF. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGF Inhibitor, Flt211, NITVTLKKFPL, VEGFR Tyrosine Kinase Inhibitor X, VEGFR1 Kinase Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H104N14O15
Molecular Weight:
1273.56
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

acetonitrile: 20%
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

An 11-amino acid peptide derived from the second Ig-like domain of vascular endothelial growth factor receptor-1 (VEGFR-1 or Flt-1) that is shown to inhibit angiogenesis in chick chorioallantoic membrane and inhibits VEGF-induced vascular permeability. Neither binds to vascular endothelial growth factor (VEGF) nor inhibits its binding to VEGFR. Reported to form a stable β-sheet structure and exists as a dimer in solution.

Biochem/physiol Actions

Cell permeable: no
Primary Target
VEGF
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Asn-Ile-Thr-Val-Thr-Leu-Lys-Lys-Phe-Pro-Leu-OH

Other Notes

Tan, D.C., et al. 2001. FEBS Lett.494, 150.

Legal Information

Sold under license of U.S. Patent 6,200,954.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service