Skip to Content
MilliporeSigma

670105

UDP-α-D-N-Acetylgalactosamine, Disodium Salt

Useful in synthesis of aryl azide derivatives that can be used in affinity labeling of glycosyltransferases and UDP-HexNAc pyrophosphorylase.

Synonym(s):

UDP-α-D-N-Acetylgalactosamine, Disodium Salt, UDP-GalNAc, 2Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H25N3O17P2 · 2Na
CAS Number:
Molecular Weight:
651.32
UNSPSC Code:
12352201
NACRES:
NA.51
MDL number:
Form:
solid
Assay:
≥97% (HPLC)
Solubility:
water: 50 mg/mL
Color:
white
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])(O[P](=O)([O-])OC[C@H]2OC([C@@H]([C@@H]2O)O)N3C=CC(=O)NC3=O)OC1O[C@@H]([C@@H]([C@@H]([C@H]1NC(=O)C)O)O)CO

InChI

1S/C17H27N3O17P2.2Na/c1-6(22)18-10-13(26)11(24)7(4-21)35-16(10)36-39(31,32)37-38(29,30)33-5-8-12(25)14(27)15(34-8)20-3-2-9(23)19-17(20)28;;/h2-3,7-8,10-16,21,24-27H,4-5H2,1H3,(H,18,22)(H,29,30)(H,31,32)(H,19,23,28);;/q;2*+1/p-2/t7-,8-,10-,11+,12-,13-,14-,15?,16?;;/m1../s1

InChI key

HXWKMJZFIJNGES-QCVFHWOISA-L

General description

Donor substrate for N-acetylgalactosaminyltransferases. A major terminal sugar of the cyst wall of Giardia lamblia. Useful in synthesis of aryl azide derivatives that can be used in affinity labeling of glycosyltransferases and UDP-HexNAc pyrophosphorylase.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Kitagawa, H., et al. 1999. Glycobiology 9, 697.
Das, S., and Gillin, F.D. 1996. Exp. Parasitol. 83, 19.
Martina, J.A., and Maccioni, H.J. 1996. J. Neurosci. Res. 46, 485.
Zeng, Y., et al. 1996. Anal. Biochem. 239, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library