Skip to Content
MilliporeSigma

Skip To

655207

TWEEN® 80

protein grade detergent, 10% solution, sterile-filtered, for immunoprecipitation and for the solubilization of membrane-bound proteins

TWEEN® 80

Synonym(s):

TWEEN® 80, Polysorbate 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 7582

Quality Segment

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

58

impurities

≤200 μM Aldehyde, ≤200 μM peroxide

color

clear colorless

CMC

12 μM (HLB number 15)

density

1.07 g/cm3 at 25 °C

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

General description

Conductivity:≤50 µmhos.
Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins (aggregation number: 58; conductivity: <50 µmhos; CMC: 12 µM; HLB number: 15). Supplied as a 10% (W/W) solution of PROTEIN GRADE TWEEN 80 detergent.

Application


  • Performance of β-carotene-loaded nanostructured lipid carriers under dynamic in vitro digestion system: Influence of the emulsifier type.: This paper discusses the performance of β-carotene-loaded nanostructured lipid carriers, where TWEEN 80 is utilized as an emulsifier. The study highlights its impact on the stability and bioaccessibility of β-carotene during digestion, offering valuable insights for food science and nutraceuticals applications (Lüdtke FL et al., 2024).

  • Fabrication and characterization of transdermal delivery of ribociclib nanoemulgel in breast cancer treatment.: This study explores the use of TWEEN 80 in developing a transdermal nanoemulgel for the delivery of ribociclib, enhancing its therapeutic efficacy against breast cancer. The work focuses on innovative drug delivery systems that improve patient compliance and drug stability (Makeen HA et al., 2024).

  • A novel antifungal nanoemulsion based on reuterin-assisted synergistic essential oils: Preparation and in vitro/in vivo characterization.: Here, TWEEN 80 is used to create a potent antifungal nanoemulsion, incorporating reuterin and essential oils. The study emphasizes the potential of nanoemulsions to significantly enhance the antifungal activity of natural compounds, promising applications in food preservation and safety (Nikkhah M et al., 2024).

  • Development and characterization of edible and active coating based on xanthan gum nanoemulsion incorporating betel leaf extract for fresh produce preservation.: This investigation uses TWEEN 80 in a xanthan gum-based nanoemulsion, integrating betel leaf extract for the preservation of fresh produce. It highlights the role of such nanoemulsions in extending the shelf life of perishable foods, with significant implications for the food industry (Aayush K et al., 2024).

Packaging

Packaged under inert gas

Physical form

Supplied as a 10% (W/W) aqueous solution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P5188P6474P6349
form

liquid

form

syrup

form

viscous liquid

form

viscous liquid

aggregation number

58

aggregation number

-

aggregation number

60

aggregation number

-

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

storage temp.

2-8°C

density

1.07 g/cm3 at 25 °C

density

1.06 g/mL at 25 °C

density

1.06 g/mL at 25 °C

density

1.07 g/cm3 at 25 °C (lit.)

impurities

≤200 μM Aldehyde, ≤200 μM peroxide

impurities

-

impurities

≤0.5 μmol/g Peroxides, ≤1.0 μmol/g Carbonyls, ≤3.0% water

impurities

≤0.5 μmol/g Peroxides, ≤1.0 μmol/g Carbonyls, ≤3.0% water

Quality Level

100

Quality Level

200

Quality Level

300

Quality Level

300


Still not finding the right product?

Explore all of our products under TWEEN® 80


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

>235.4 °F - closed cup

flash_point_c

> 113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review