Skip to Content
MilliporeSigma

650357

Trypsin Inhibitor, Soybean, High Activity

Trypsin Inhibitor, Soybean, High Activity, CAS 9035-81-8 is of the several protease inhibitors in soybeans. Inhibits trypsin in 1:1 stoichiometry.

Synonym(s):

Trypsin Inhibitor, Soybean, High Activity

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
NACRES:
NA.28
UNSPSC Code:
12352202
Assay:
≥90% (SDS-PAGE)
Form:
lyophilized solid
Storage condition:
OK to freeze, desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (SDS-PAGE)

form

lyophilized solid

specific activity

≥7,000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

white

solubility

67 mM potassium phosphate buffer, pH 7.6: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Component VI of Rackis. Does not inhibit chymotrypsin activity.
Inactivates trypsin on an equal molar basis. To ensure complete inhibition of trypsin activity, a working concentration of soybean trypsin inhibitor should exceed that of trypsin by a factor of at least 2.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Trypsin
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

One unit is defined as the amount of protein that will inhibit 1 unit of trypsin activity using BAEE as a substrate at 25°C, pH 7.6.
Uchino, T., et al. 1993. J. Biol. Chem.268, 527.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

11 - Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
650357-250MG04055977184532
650357-100MG04055977184525