Skip to Content
MilliporeSigma

648468

Triton™ X-114 Solution

10% in H₂O, PROTEIN GRADE®

Synonym(s):

(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol solution, t-Octylphenoxypolyethoxyethanol; Polyethylene glycol tert-octylphenyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.21
UNSPSC Code:
12161902
MDL number:
Beilstein/REAXYS Number:
2315025
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Triton™ X-114, PROTEIN GRADE® Detergent, 10% Solution, Sterile-Filtered, Non-ionic detergent and emulsifier. HLB 12.4. Cloud point 22°C.

Quality Segment

description

RTECS - MD0907600

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<200 μM Aldehyde, <200 μM peroxide

color

clear

conductivity

<50 μmho

CMC

350 μM

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

General description

HLB 12.4. Cloud point 22°C.
Non–ionic detergent and emulsifier.
Non-ionic detergent and emulsifier (cloud point: 22°C; CMC: 350 µM; HLB: 12.4).

Application


  • Determination of nine aromatic amines in water by cloud point extraction-gas chromatography-mass spectrometry.: This study utilized Triton X-114 for cloud point extraction to enhance the detection sensitivity of aromatic amines in water samples. The detergent played a crucial role in the efficient extraction and subsequent analysis of these compounds, highlighting its utility in environmental monitoring and analytical chemistry (Yang et al., 2024).

  • An innovative lab-scale production for a novel therapeutic DNA vaccine candidate against rheumatoid arthritis.: Triton X-114 was employed in the purification steps to ensure the integrity and activity of the vaccine components. This detergent′s role in protein solubilization and membrane protein isolation was pivotal in the development of the therapeutic candidate, demonstrating its importance in biotechnological applications (Long et al., 2024).

  • Lipoprotein signal peptidase-deficient Streptococcus pneumoniae exhibits impaired Toll-like receptor 2-stimulatory activity.: The study used Triton X-114 for the extraction and analysis of membrane proteins, which facilitated the investigation of bacterial interactions with host immune receptors. This application underscores the detergent′s effectiveness in microbial research and immunology (Domon et al., 2024).

  • Improvement in the Chromium(VI)-Diphenylcarbazide Determination Using Cloud Point Microextraction; Speciation of Chromium at Low Levels in Water Samples.: Triton X-114 was used for cloud point extraction to improve the sensitivity and accuracy of chromium speciation in water samples. This method highlights the detergent′s role in environmental analytical chemistry, enhancing the detection of trace metal species (Mouco-Novegil et al., 2023).

Packaging

Packaged under inert gas

Physical form

A 10% aqueous solution.

Other Notes

D’Arrigo, A., et al. 1993. J. Biol. Chem.268, 2802.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Disclaimer

Toxicity: Irritant (B)


Pictograms

CorrosionEnvironment

Signal Word

Danger

Storage Class

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1 - Skin Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library