Skip to Content
MilliporeSigma
All Photos(1)

Documents

647926-M

Sigma-Aldrich

Trichostatin A, Streptomyces sp.

InSolution, ≥98%, reversible inhibitor of histone deacetylase

Synonym(s):

InSolution Trichostatin A, Streptomyces sp., 4,6-Dimethyl-7-[p-dimethylaminophenyl]-7-oxahepta-2,4-dienohydroxamic Acid, HDAC Inhibitor IX, TSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O3
Molecular Weight:
302.37
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

storage temp.

−20°C

General description

A potent and reversible inhibitor of histone deacetylase. Blocks cell cycle progression at the G1 phase in HeLa cells and induces a 12-fold increase in intracellular levels of gelsolin. Induces reversion of oncogenic ras-tranformed NIH 3T3 cells to a normal morphology. Inhibits IL-2 gene expression (IC50 = 73 nM) in Jurkat cells and shows immunosuppressive activity in a mouse model.

Biochem/physiol Actions

Primary Target
histone deactylase
Target IC50: 73 nM inhibiting IL-2 gene expression in Jurkat cells

Warning

Toxicity: Harmful (C)

Physical form

A 10 mM (500 µg/165 µl) solution of Trichostatin A, Streptomyces sp. (Cat. No. 647925) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service