Skip to Content
MilliporeSigma
All Photos(1)

Documents

6210-OP

Sigma-Aldrich

Methylene Blue Trihydrate

OmniPur® Grade

Synonym(s):

Methylene Blue trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S · 3H2O
CAS Number:
Molecular Weight:
373.90
Beilstein:
3599847
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

iridescent green-black

mp

190 °C (dec.) (lit.)

shipped in

ambient

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;/h5-10H,1-4H3;1H/q+1;/p-1

InChI key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Post-electrophoretic stain for DNA. Stains RNA in bacterialogical procedures. Non-intercalating. Molecular biology grade.

Specifications:
Arsenic (As): ≤0.0008%
Ash: ≤1.0%
Copper (Cu): ≤0.02%
Lead (Pb): ≤0.001%
Moisture (H2O): ≤15.0%
Molar absorptivity Em (661 nm, water): ≥69000
Zinc (Zn): ≤0.001%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service