Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616451-M

Sigma-Aldrich

TGF-β RI Kinase Inhibitor

The TGF-β RI Kinase Inhibitor, also referenced under CAS 396129-53-6, controls the biological activity of TGF-β RI Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

TGF-β RI Kinase Inhibitor, Transforming Growth Factor-β Type I Receptor Kinase Inhibitor, ALK5 Inhibitor I, [3-(Pyridin-2-yl)-4-(4-quinonyl)]-1H-pyrazole, TβR-I Inhibitor, LY-364947, HTS-466284, [3-(Pyridin-2-yl)-4-(4-quinonyl)]-1H-pyrazole, TβR-I Inhibitor, Transforming Growth Factor-β Type I Receptor Kinase Inhibitor, ALK5 Inhibitor I, LY-364947, HTS-466284

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12N4
CAS Number:
Molecular Weight:
272.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

SMILES string

[nH]1ncc(c1c4ncccc4)c2c3c(ncc2)cccc3

InChI

1S/C17H12N4/c1-2-6-15-13(5-1)12(8-10-19-15)14-11-20-21-17(14)16-7-3-4-9-18-16/h1-11H,(H,20,21)

InChI key

IBCXZJCWDGCXQT-UHFFFAOYSA-N

General description

A cell-permeable diheteroaryl-substituted pyrazole compound that acts as a potent, selective, reversible, and ATP-competitive inhibitor of TGF-β Receptor I kinase (IC50 = 51 nM). Displays ~15-fold greater selectivity over p38α MAP kinase (IC50 = 740 nM). Shown to inhibit TGF-β-dependent cellular growth (IC50 = 89 nM in NIH 3T3 mouse fibroblasts) and transcription activation (IC50 = 47 nM in mink lung cells).
A cell-permeable, a potent, selective, reversible, and ATP-competitive inhibitor of TGF-b RI serine/threonine kinase (IC50 = 51 nM). Displays ~15-fold greater selectivity over p38α MAP kinase (IC50 = 740 nM). Reported to inhibit TGF-b-dependent cellular growth (IC50 = 89 nM in mouse NIH3T3 fibroblasts) and transcription activation (IC50 = 47 nM in mink lung cells).

Biochem/physiol Actions

Primary Target
TGF-β receptor I kinase
Target IC50: 51 nM against TGF-β Receptor I kinase; 89 nM against TGF-β-dependent cellular growth in NIH 3T3 mouse fibroblasts; 47 nM against TGF-β-dependent transcription activation in mink lung cells

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sawyer, J.S., et al. 2003. J. Med. Chem.46, 3953.
Singh, J., et al. 2003. Bioorg. Med. Chem. Lett.13, 4355.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service