Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616373

Sigma-Aldrich

Tpl2 Kinase Inhibitor

The Tpl2 Kinase Inhibitor, also referenced under CAS 871307-18-5, controls the biological activity of Tpl2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Tpl2 Kinase Inhibitor, 4-(3-Chloro-4-fluorophenylamino)-6-(pyridin-3-yl-methylamino)-3-cyano-[1,7]-naphthyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14ClFN6
CAS Number:
Molecular Weight:
404.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1c(cc(cc1)Nc2c3c(ncc2C#N)cnc(c3)NCc4cnccc4)Cl

InChI

1S/C21H14ClFN6/c22-17-6-15(3-4-18(17)23)29-21-14(8-24)11-26-19-12-28-20(7-16(19)21)27-10-13-2-1-5-25-9-13/h1-7,9,11-12H,10H2,(H,26,29)(H,27,28)

InChI key

NMEUKWOOQOHUNA-UHFFFAOYSA-N

General description

A cell-permeable naphthyridine compound that acts as a potent, reversible, and ATP-competitive inhibitor of Tpl2 kinase (IC50 = 50 nM). Displays significant selectivity over other related kinases (IC50 = 5, >40, 110, 180, >400 and >400 µM for EGFR, MEK, MK2, p38, Src, and PKC, respectively). Shown to inhibit LPS-induced TNF-α production both from primary human monocytes and in whole blood (IC50 = 700 nM and 8.5 µM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Tpl2 Kinase
Product competes with ATP.
Reversible: yes
Target IC50: 50 nM against Tpl2 kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gavrin, L.K., et al. 2005. Bioorg. Med. Chem. Lett.15, 5288.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kara D Wyatt et al.
PloS one, 17(1), e0262832-e0262832 (2022-01-21)
Tumor progression locus 2 (Tpl2) is a serine/threonine kinase that regulates the expression of inflammatory mediators in response to Toll-like receptors (TLR) and cytokine receptors. Global ablation of Tpl2 leads to severe disease in response to influenza A virus (IAV)
Cory M Johannessen et al.
Nature, 468(7326), 968-972 (2010-11-26)
Oncogenic mutations in the serine/threonine kinase B-RAF (also known as BRAF) are found in 50-70% of malignant melanomas. Pre-clinical studies have demonstrated that the B-RAF(V600E) mutation predicts a dependency on the mitogen-activated protein kinase (MAPK) signalling cascade in melanoma-an observation
Aikaterini Nanou et al.
Cell reports, 35(8), 109168-109168 (2021-05-27)
Increased vascular permeability and leakage are hallmarks of several pathologies and determine disease progression and severity by facilitating inflammatory/metastatic cell infiltration. Using tissue-specific genetic ablation in endothelial cells, we have investigated in vivo the role of Tumor progression locus 2 (Tpl2)
Timothy M Errington et al.
eLife, 10 (2021-12-08)
As part of the Reproducibility Project: Cancer Biology, we published Registered Reports that described how we intended to replicate selected experiments from 29 high-impact preclinical cancer biology papers published between 2010 and 2012. Replication experiments were completed and Replication Studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service