Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

614853

Sigma-Aldrich

Topoisomerase II Inhibitor, BNS-22

The Topoisomerase II Inhibitor, BNS-22 controls the biological activity of Topoisomerase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Topoisomerase II Inhibitor, BNS-22, BNS-22

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25NO5
Molecular Weight:
407.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

N3(CCCc4c3cccc4)C(=O)c1c2c(c(cc1OC)OC)C(=CC(=O)O2)CCC

InChI

1S/C24H25NO5/c1-4-8-16-13-20(26)30-23-21(16)18(28-2)14-19(29-3)22(23)24(27)25-12-7-10-15-9-5-6-11-17(15)25/h5-6,9,11,13-14H,4,7-8,10,12H2,1-3H3

InChI key

LRPUQCTZOSTSGL-UHFFFAOYSA-N

General description

A potent, 2H-chromen-2-one small molecule inhibitor that is selective for TOP2 (IC50 = 2.8 µM and 0.42 µM for TOP2α and TOP2β, respectively). Unlike TOP2 poisons such as etoposide, it does not lead to DNA damage associated with DNA Double-Strand Breaks, as evidenced by the lack of γ-H2AX accumulation but instead inhibits kinetoplast DNA decatenation in vitro. This compound affects cell cycle at the M phase and disturbs mitotic spindle formation in HeLa cells at 3-10 µM. It also exhibits significant anti-proliferative activities against several human cancer cell lines.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kawatani, M., et al. 2011. Chem Biol.18, 743.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arun Pal et al.
Life science alliance, 4(4) (2021-02-24)
Intronic hexanucleotide repeat expansions (HREs) in C9ORF72 are the most frequent genetic cause of amyotrophic lateral sclerosis, a devastating, incurable motoneuron (MN) disease. The mechanism by which HREs trigger pathogenesis remains elusive. The discovery of repeat-associated non-ATG (RAN) translation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service