Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

613810

Sigma-Aldrich

TNAP Inhibitor

The TNAP Inhibitor, also referenced under CAS 496014-13-2, controls the biological activity of TNAP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

TNAP Inhibitor, 2,5-Dimethoxy-N-(quinolin-3-yl)benzenesulfonamide, Tissue-Nonspecific Alkaline Phosphatase Inhibitor, MLS-0038949, MLS-0038949, 2,5-Dimethoxy-N-(quinolin-3-yl)benzenesulfonamide, Tissue-Nonspecific Alkaline Phosphatase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O4S
CAS Number:
Molecular Weight:
344.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc2cnc3c(c2)cccc3)c1c(ccc(c1)OC)OC

InChI

1S/C17H16N2O4S/c1-22-14-7-8-16(23-2)17(10-14)24(20,21)19-13-9-12-5-3-4-6-15(12)18-11-13/h3-11,19H,1-2H3

InChI key

JZSPHTPWUXNQGB-UHFFFAOYSA-N

General description

A cell-permeable quinolinyl-benzenesulfonamide compound that acts as a potent reversible inhibitor against tissue-nonspecific (TN) alkaline phosphatase (AP) activity (IC50 = 190 nM) with CYP2C19 being the only other known target, while exhibiting little or no inhibitory activity in over 250 assays involving other enzymes, including, but not limited to, placental and intestinal APs (IC50 >100 µM), PHOSPHO1 phosphatase, and NPP1 phosphodiesterase. Kinetic studies indicate that the inhibition is uncompetitive with respect to the phosphate donor substrate and noncompetitive with respect to the acceptor substrate, indicating an allosteric inhibition mechanism. Subcutaneous dosing in rats in vivo is reported to result in plasma inhibitor concentrations in the therapeutic range (Cmax = 1.5 µg/ml 1 hr after 4.13 mg/kg by s.c.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and store at (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Dahl, R., et al. 2009. J. Med. Chem.52, 6919.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony W Tolcher et al.
Journal for immunotherapy of cancer, 11(2) (2023-02-07)
Cluster of differentiation (CD)73-adenosine and transforming growth factor (TGF)-β pathways are involved in abrogated antitumor immune responses and can lead to protumor conditions. This Phase 1 study (NCT03954704) evaluated the safety, pharmacokinetics, pharmacodynamics, and efficacy of dalutrafusp alfa (also known
Andrew C Yang et al.
Nature, 583(7816), 425-430 (2020-07-03)
The vascular interface of the brain, known as the blood-brain barrier (BBB), is understood to maintain brain function in part via its low transcellular permeability1-3. Yet, recent studies have demonstrated that brain ageing is sensitive to circulatory proteins4,5. Thus, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service