Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

612200-M

Sigma-Aldrich

Tissue Plasminogen Activator, Human

Synonym(s):

tPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

specific activity

≥300 KU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage temp.

−20°C

General description

Native tissue plasminogen activator from a human melanoma cell line. A serine protease that contains a single chain of 527 amino acid residues. Binds to fibrin via lysine binding sites at its amino terminus and activates bound plasminogen. Cleaves plasminogen to form active plasmin, the proteolytic enzyme that dissolves clots. Reported to be involved in neurite outgrowth, regeneration, and migration. Plays an important role in excitotoxin-induced neuronal degeneration. Dihydropyridine Ca2+ antagonist may increase levels of tPA.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 100 mM PBS, 3.5 mg/ml L-arginine, 0.001% Tween®-80 Detergent.

Preparation Note

Prepared from cell line that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Reconstitute with diH₂O to a final concentration of 1 mg/ml (e.g., 100 µg should be reconstituted in 100 µl).

Other Notes

Kim, Y.H., et al. 1999. Science284, 647.
Tsirka, S.E., et al. 1995. Nature 377, 340.
Winther, K., et al. 1992. J. Cardiovasc. Pharmacol.19, S21.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service