Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

609315-M

Millipore

Thyroid Hormone Receptor Antagonist, 1-850

The Thyroid Hormone Receptor Antagonist, 1-850, also referenced under CAS 251310-57-3, controls the biological activity of Thyroid Hormone Receptor. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

Thyroid Hormone Receptor Antagonist, 1-850, TR Antagonist, 1-850, 2-(2-(-(4-Nitrophenyl)-4-piperidinylidene)acetyl-N-(3-(trifluoromethyl)phenyl)-1-hydrazine Carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20F3N5O4
CAS Number:
Molecular Weight:
463.41
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

General description

A cell-permeable hydrazinyl-carboxamide compound that acts as a selective and high-affinity thyroid hormone receptor (TR) antagonist (IC50 = 1.5 µM in HeLa cells). Shown to block T3- (L-triiodothyronine) mediated interaction of TRα with NRC (nuclear receptor coactivator) and prevent the stimulation of gene expression. Exerts no effect on the retinoic acid receptor α (RARα) activity.
A cell-permeable, selective, high-affinity thyroid hormone receptor (TR) antagonist (IC50 = 1.5 µM in HeLa cells). Reported to block T3-L-triiodothyronine)-mediated interaction of TRα with nuclear receptor coactivator (NRC) and prevent stimulation of gene expression. Exhibits no antagonistic effects on retinoic acid receptor α (RARα) activity.

Biochem/physiol Actions

Primary Target
Thyroid Hormone Receptor
Target IC50: 1.5 µM as antagonist for thyroid hormone receptor (TR) in HeLa cells

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schapira, M., et al. 2003. Proc. Natl. Acad. Sci. USA100, 7354.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service