Skip to Content

6060

Thymidine

Synonym(s):

Thymidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
UNSPSC Code:
12352208
NACRES:
NA.51
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Solubility:
water: 10 mg/mL
Storage temp.:
10-30°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - XP2071000
Merck USA index - 14, 9397

Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 10 mg/mL

cation traces

heavy metals: ≤10 ppm

shipped in

ambient

storage temp.

10-30°C

SMILES string

[nH]1[c]([n](cc([c]1=O)C)C2OC(C(C2)O)CO)=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)

InChI key

IQFYYKKMVGJFEH-UHFFFAOYSA-N

General description

Used in conjunction with hypoxanthine and aminopterin for the selection of hybridoma cell lines. Also reported to induce cell death in L-cells.

Preparation Note

Following reconstitution, sterilize by filtration using a 0.22 µm filter, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Amenta, J.S., et al. 1993. In Vitro Cell Dev. Biol. Anim.29A, 855.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Thymidine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library