Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
UNSPSC Code:
12191601
NACRES:
NA.22
MDL number:
Beilstein/REAXYS Number:
1098229
Assay:
99.8%
Grade:
anhydrous
Bp:
64.7 °C (lit.)
Vapor pressure:
410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
Product Name
Methanol, anhydrous, 99.8%
grade
anhydrous
Quality Segment
vapor density
1.11 (vs air)
vapor pressure
410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
assay
99.8%
autoignition temp.
725 °F
expl. lim.
36 %
refractive index
n20/D 1.329 (lit.)
bp
64.7 °C (lit.)
mp
−98 °C (lit.)
density
0.791 g/mL at 25 °C (lit.)
format
neat
SMILES string
CO
InChI
1S/CH4O/c1-2/h2H,1H3
InChI key
OKKJLVBELUTLKV-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Methanol
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
target_organs
Eyes,Central nervous system
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 5895962000 | 04065267634609 |
| 5895961000 | 04065267633596 |
| 5895960100 | 04065267635637 |
| 5895960250 | 04065267633589 |


