Skip to Content
MilliporeSigma

5.89581

Dichloromethane

contains 15-150 ppm amylene as stabilizer, anhydrous, 99.8%

Synonym(s):

Methylene chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH2Cl2
CAS Number:
Molecular Weight:
84.93
MDL number:
NACRES:
NA.22
UNSPSC Code:
12352101
Beilstein/REAXYS Number:
1730800
Assay:
99.8%
Grade:
anhydrous
Bp:
39.8-40 °C (lit.)
Vapor pressure:
24.45 psi ( 55 °C), 6.83 psi ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

anhydrous

Quality Segment

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C), 6.83 psi ( 20 °C)

assay

99.8%

autoignition temp.

1223 °F

contains

15-150 ppm amylene as stabilizer

expl. lim.

22 %

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

General description

Our anhydrous solvents deliver exceptionally low water content, making them the ideal choice for moisture-sensitive laboratory applications. Packaged in the exclusive Sure/Seal™ septum bottle system, they are reliably protected against moisture ingress from first use to last drop. The innovative Sure/Seal™ closure combines an advanced plug-style liner with an airtight elastomer and crimp cap design to ensure consistent product integrity. Three unique liner materials provide optimal chemical compatibility across a broad range of solvents. Sure/Seal™ packaging is proven to maintain anhydrous conditions even after repeated punctures. With more than 70 products across all major solvent families, available in pack sizes from 100 mL to 2 L, the portfolio meets virtually every laboratory need. By choosing ready-to-use anhydrous solvents in Sure/Seal™ packaging, researchers eliminate the need for time-consuming and hazardous in-house solvent drying and distillation - saving time, reducing risk, and improving reproducibility.

Application

Anhydrous solvents in Sure/Seal™ packaging are essential wherever water contamination would compromise experimental results. They are widely used in moisture-sensitive applications such as organic synthesis - including Grignard reactions, organometallic chemistry, and catalysis - as well as in peptide, oligonucleotide, and pharmaceutical synthesis.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?

Explore all of our products under Dichloromethane


Signal Word

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Pictograms

Health hazardExclamation mark

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library