Skip to Content
MilliporeSigma

586006

Thapsigargin

≥97% (TLC), liquid, endoplasmic reticular Ca2+-ATPase inhibitor, Calbiochem®

Synonym(s):

InSolution Thapsigargin, (3 S,3a R,4 S,6 S,6a R,7 S,8 S,9b S)-6-Acetoxy-4-(butyryloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-((Z)-2-methylbut-2-enoyloxy)-2-oxo-2,3,3a,4,5,6,6a,7,8,9b-decahydroazuleno[4,5-b]furan-7-yl octanoate, (3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-Acetoxy-4-(butyryloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-((Z)-2-methylbut-2-enoyloxy)-2-oxo-2,3,3a,4,5,6,6a,7,8,9b-decahydroazuleno[4,5-b]furan-7-yl octanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H50O12
CAS Number:
Molecular Weight:
650.75
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (TLC)
Form:
liquid
Storage condition:
OK to freeze, avoid repeated freeze/thaw cycles, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Thapsigargin, InSolution, ≥97%, tumor-promoting sesquiterpene lactone

Quality Segment

assay

≥97% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles, desiccated (hygroscopic), protect from light

shipped in

dry ice

storage temp.

−70°C

SMILES string

O1[C@@H]2[C@]([C@H](C[C@@]([C@H]3[C@@H]([C@H](C(=C32)C)OC(=O)\C(=C/C)\C)OC(=O)CCCCCCC)(OC(=O)C)C)OC(=O)CCC)([C@@](C1=O)(O)C)O

InChI

1S/C34H50O12/c1-9-12-13-14-15-17-24(37)43-28-26-25(20(5)27(28)44-30(38)19(4)11-3)29-34(41,33(8,40)31(39)45-29)22(42-23(36)16-10-2)18-32(26,7)46-21(6)35/h11,22,26-29,40-41H,9-10,12-18H2,1-8H3/b19-11-/t22-,26+,27-,28-,29-,32-,33+,34+/m0/s1

InChI key

IXFPJGBNCFXKPI-FSIHEZPISA-N

General description

A cell-permeable tumor-promoting sesquiterpene lactone that releases Ca2+ by inhibiting endoplasmic reticular Ca2+-ATPase (IC50 = 4 - 13 nM). Does not increase inositol phosphates and has no significant effect on protein kinase C. Increases Ca2+-dependent Na+ influx in human platelets in a dose-dependent manner. Activates apoptosis in rat thymocytes and induces apoptosis in human hepatoma cells.Specifically inhibits the fusion of autophagosomes with lysosomes, and induces autophagy.

Biochem/physiol Actions

Cell permeable: yes
Reversible: no

Packaging

Packaged under inert gas

Physical form

A 10 mM (2 mg/307 µL) solution of Thapsigargin (Cat. No. 586005) in DMSO.

Other Notes

Won, J.G. and Orth, D.N. 1995. Endocrinology136, 5399.Tepel, M., et al. 1994. Biochim. Biophys. Acta1220, 248.Begum, N., et al. 1993. J. Biol. Chem.268, 3352.Low, A.M., et al. 1993. Eur. J. Pharmacol.230, 53.Tsukamoto, A. and Karneko, Y. 1993. Cell. Biol. Int.17, 969.Wong, W.L., et al. 1993. Biochem. J.289, 71.Glennon, M.C., et al. 1992. J. Biol. Chem.267, 25568.Xu, Y., et al. 1992. J. Neurochem.59, 2224.Mason, M.J., et al. 1991. J. Biol. Chem.266, 20856.Thastrup, O., et al. 1990. Proc. Natl. Acad. Sci. USA87, 2466.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Still not finding the right product?

Explore all of our products under Thapsigargin


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library