Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58346-M

Sigma-Aldrich

Tetracycline, Hydrochloride

CAS 64-17-5 is a broad-based antibiotic agent that blocks protein synthesis by inhibiting binding of aminoacyl tRNA to the A-site of ribosomes. Induces cold shock-response and enhances P450 expression in bacteria.

Synonym(s):

Tetracycline, Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8
CAS Number:
Molecular Weight:
444.43
MDL number:
UNSPSC Code:
51284043

Quality Level

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

concentration

≥900 μg/mg

color

yellow

solubility

water: 20 mg/mL
ethanol: 5 mg/mL

storage temp.

10-30°C

InChI

1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25-26,30-32H,7,23H2,1-3H3;1H/b20-14-;/t9-,10-,15-,21+,22-;/m0./s1

InChI key

DYQBMRZOOXYHIM-NCKOGSTASA-N

General description

An antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing binding of aminoacyl tRNA to the A-site and blocking translocation. Used commonly for selection of the bacterial tetr gene whose product, a 399-amino acid membrane-associated protein, functions in active export of the drug from the cell.
Broad-based antibiotic agent that blocks protein synthesis by inhibiting binding of aminoacyl tRNA to the A-site of ribosomes. Induces cold shock-response and enhances P450 expression in bacteria.

Biochem/physiol Actions

Primary Target
binding of aminoacyl tRNA to the A-site of ribosomes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, sterilize by filtration through a 0.22 µm pore-size filter, aliquot and freeze (-20°C). Stock solutions are stable for 1 month at -20°C. Agar plates are stable for 2 weeks at 4°C. Autoclaved media should be cooled to ≤55°C before adding tetracycline.

Other Notes

Kusano, K., et al. 1999. Arch. Biochem. Biophys. 367, 129.
Lech, K. and Brent, R. 1995. In Current Protocols in Molecular Biology, Vol. 1 (Ausubel, F.M., et al. eds.) John Wiley & Sons, New York Unit 1.4.
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition. Cold Spring Harbor, New York, p. 1.6, A.6.
Chopra, I. and Howe, T.G.B. 1978. Microbiol. Rev. 42, 707.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service