Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

575003-M

Sigma-Aldrich

α-Synuclein A53T, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

Recombinant, human α-synuclein containing an A53T mutation and expressed in E. coli. α-synuclein is an acidic neuronal protein of 140 amino acids that has been implicated in the pathogenesis of Parkinson′s disease and related neurodegenerative disorders. α-Synuclein (A53T) is a point mutant of the α-synuclein gene that has been linked to autosomal dominant early onset Parkinson′s Disease (PD). The apparent molecular weight on SDS-PAGE is ~19-20 kDa.
Recombinant, human α-synuclein containing an A53T mutation and expressed in E. coli. A point mutant (A53T) of α-synuclein gene that has been linked to autosomal dominant early-onset Parkinson’s disease (PD).

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 20 mM Tris-HCl, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Lashuel, H.A., et al. 2002. J. Mol. Biol.322, 1089.
Park, S.M., et al. 2002. Blood100, 2506.
Polymeropoulos, M.H., et al. 1997. Science276, 2045.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service