Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

573102

Sigma-Aldrich

S3I-201

≥95% (HPLC), solid, STAT3 inhibitor, Calbiochem®

Synonym(s):

STAT3 Inhibitor VI, S3I-201, 2-Hydroxy-4-(((4-methylphenyl)sulfonyloxy)acetyl)amino)-benzoic acid, NSC 74859

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15NO7S
CAS Number:
Molecular Weight:
365.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

STAT3 Inhibitor VI, S3I-201, STAT3 Inhibitor VI, S3I-201, CAS 501919-59-1, is a cell-permeable compound that binds Stat3-SH2 domain and blocks Stat3 phosphorylation, dimerization, DNA-binding, & Stat3-dependent transcription.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 10 mg/mL (Use only fresh, anhydrous DMSO.)

shipped in

ambient

storage temp.

−20°C

SMILES string

[S](=O)(=O)(OCC(=O)Nc2cc(c(cc2)C(=O)O)O)c1ccc(cc1)C

InChI

1S/C16H15NO7S/c1-10-2-5-12(6-3-10)25(22,23)24-9-15(19)17-11-4-7-13(16(20)21)14(18)8-11/h2-8,18H,9H2,1H3,(H,17,19)(H,20,21)

InChI key

HWNUSGNZBAISFM-UHFFFAOYSA-N

General description

A cell-permeable amidosalicylic acid compound that binds Stat3-SH2 domain and prevents Stat3 phosphorylation/activation, dimerization, DNA-binding, and Stat3-dependent transcription. Shown to arrest Stat3-dependent tumor growth both in cultures in vitro (effective conc. ≤100 µM) and in a murine xenograft model in vivo (5 mg/kg, i.v.). Also available as a 50 mM solution in DMSO (Cat. No. 573130).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Supriya Murthy et al.
Cell death & disease, 13(1), 53-53 (2022-01-14)
The danger signal extracellular calcium is pathophysiologically increased in the synovial fluid of patients with rheumatoid arthritis (RA). Calcium activates the NLRP3-inflammasome via the calcium-sensing receptor in monocytes/macrophages primed by lipopolysaccharide, and this effect is mediated by the uptake of
Mutation-Driven S100A8 Overexpression Confers Aberrant Phenotypes in Type 1 CALR-Mutated MPN.
Wang, et al.
International Journal of Molecular Sciences, 24 (2023)
Kentaro Kajiwara et al.
Life science alliance, 4(4) (2021-02-13)
Compensatory growth of organs after loss of their mass and/or function is controlled by hepatocyte growth factor (HGF), but the underlying regulatory mechanisms remain elusive. Here, we show that CUB domain-containing protein 1 (CDCP1) promotes HGF-induced compensatory renal growth. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service