Skip to Content
MilliporeSigma
All Photos(1)

Documents

572631

Sigma-Aldrich

SU5402

InSolution, ≥95%, Inhibits the tyrosine kinase activity of FGFR1

Synonym(s):

InSolution SU5402

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

General description

Inhibits the tyrosine kinase activity of FGFR1 (IC50 = 10-20 µM in the presence of 1 mM ATP) by interacting with the catalytic domain of FGFR1. Also inhibits aFGF-induced phosphorylation of ERK1 and ERK2 (IC50 = 10-20 µM). In contrast to SU4984, it acts only as a weak inhibitor of tyrosine phosphorylation of the PDGF receptor. Does not inhibit the phosphorylation of insulin receptor and exhibits no inhibitory effect on EGF receptor kinase.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/169 µl) solution of SU5402 (Cat. No. 572630) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Mohammadi, M., et al. 1997. Science 276, 955.

Legal Information

Not available for sale in the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lydvina Meister et al.
Development (Cambridge, England), 149(10) (2022-05-17)
The fibroblast growth factor (FGF) signalling pathway plays various roles during vertebrate embryogenesis, from mesoderm formation to brain patterning. This diversity of functions relies on the fact that vertebrates possess the largest FGF gene complement among metazoans. In the cephalochordate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service