Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567547

Millipore

Sodium phosphate dibasic

Molecular Biology Grade; Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers.

Synonym(s):

Sodium phosphate dibasic, Na₂HPO₄, Sodium Phosphate, Dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

Quality Level

Assay

≥98.5% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

water: 10%

cation traces

heavy metals (as Pb): ≤0.001%

absorption

≤0.01 at 260 nm
≤0.01 at 280 nm

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Molecular biology reagent (absorbance: ≤0.01 at 260 nm and 280 nm).
Useful in conjunction with Sodium Phosphate, Monobasic (Cat. No. 567545 or 567549) in the preparation of biological buffers.

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service