Skip to Content
MilliporeSigma

559286

RSK Inhibitor II

The RSK Inhibitor II, also referenced under CAS 501437-28-1, controls the biological activity of RSK. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

RSK Inhibitor II, 2-(3,5-Difluoro-4-hydroxy-anilino)-8-isopentyl-5,7-dimethyl-7H-pteridin-6-one, racemic, p90 Ribosomal S6 Kinase Inhibitor II, BI-D1870, BID-1870, BID1870

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H23F2N5O2
CAS Number:
Molecular Weight:
391.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(c(cc(c1)Nc2nc3c(cn2)N(C(=O)C(N3CCC(C)C)C)C)F)O

InChI

1S/C19H23F2N5O2/c1-10(2)5-6-26-11(3)18(28)25(4)15-9-22-19(24-17(15)26)23-12-7-13(20)16(27)14(21)8-12/h7-11,27H,5-6H2,1-4H3,(H,22,23,24)

InChI key

DTEKTGDVSARYDS-UHFFFAOYSA-N

General description

The racemic mixture of a cell-permeable dihydropteridinone that acts as a potent and selective RSK inhibitor (IC50 = 31, 24, 18, and 15 nM against RSK1, 2, 3, and 4, respectively; [ATP] = 100 µM) by targeting RSK N-terminal kinase domain in an ATP-competitive manner, while inhibiting 11 other kinases only at higher concentrations (IC50 ≥100 nM) and displaying little activity against 39 others. Shown to selectively inhibit RSK-dependent, but not RSK-independent, cellular protein phosphorylations in PMA-stimulated human HEK-293 cells and EGF-stimulated rat Rat-2 cells (IC50 ~1 µM). A useful pharmacological reagent for studying RSK-mediated cellular signaling events.

Packaging

Packaged under inert gas

Other Notes

Carriere, A., et al. 2008. Curr. Biol.18, 1269
Sapkota, G.P., et al. 2007. Biochem. J.401, 29.
Zaru, R., et al. 2007. Nat. Immunol.8, 1227.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library