Skip to Content
MilliporeSigma

Skip To

553525

RAD51 Inhibitor B02

The RAD51 Inhibitor, B02 controls the biological activity of RAD51. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

RAD51 Inhibitor, B02, ( E)-3-Benzyl-2-(2-(pyridin-3-yl)vinyl)quinazolin-4(3H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H17N3O
CAS Number:
Molecular Weight:
339.39
UNSPSC Code:
12352200
NACRES:
NA.28
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

RAD51 Inhibitor, B02, The RAD51 Inhibitor, B02 controls the biological activity of RAD51. This small molecule/inhibitor is primarily used for Cell Structure applications.

Quality Segment

assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]2([c](c3c(nc2\C=C\c4cnccc4)cccc3)=O)Cc1ccccc1

InChI

1S/C22H17N3O/c26-22-19-10-4-5-11-20(19)24-21(13-12-17-9-6-14-23-15-17)25(22)16-18-7-2-1-3-8-18/h1-15H,16H2/b13-12+

InChI key

GEKDQXSPTHHANP-OUKQBFOZSA-N

General description

A cell-permeable pyridinylvinyl-quinazolinone compound that is shown to specifically inhibit human RAD51 (IC50 = 27.4 µM). Does not affect RecA even at much higher concentration (~250 µM). Directly interacts with RAD51 (Kd = 5.6 µM), and disrupts its binding to DNA and nucleoprotein filament formation. Blocks double-strand break-induced homologous recombination and enhances sensitivity of cells to Cisplatin (Cat. No. 232120) and Mitomycin C (Cat. No. 47589), (Cat. No. 475820). Diminishes co-aggregate formation between RAD51-ssDNA filament and dsDNA. RAD51-Stimulatory Compound-1, RS-1 (Cat. No. 553510) is also available.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Huang, F., et al. 2012. J. Med. Chem.55, 3031.
Huang, F., et al. 2011. ACS Chem. Biol.6, 628.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0364SRP2090ABE257
description

The RAD51 Inhibitor, B02 controls the biological activity of RAD51. This small molecule/inhibitor is primarily used for Cell Structure applications.

description

≥98% (HPLC)

description

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

description

from rabbit, purified by affinity chromatography

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥80% (SDS-PAGE)

assay

-

form

powder

form

powder

form

frozen liquid

form

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−70°C

storage temp.

-

solubility

DMSO: 50 mg/mL

solubility

DMSO: ≥5 mg/mL

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under RAD51 Inhibitor, B02


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters