Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553008

Sigma-Aldrich

Raf1 Kinase Inhibitor I

The Raf1 Kinase Inhibitor I, also referenced under CAS 220904-83-6, controls the biological activity of Raf1 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Raf1 Kinase Inhibitor I, 5-Iodo-3-[(3,5-dibromo-4-hydroxyphenyl)methylene]-2-indolinone, GW 5074

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8Br2INO2
CAS Number:
Molecular Weight:
520.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥97% (HPLC)

form

solid

potency

9 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark orange

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C15H8Br2INO2/c16-11-4-7(5-12(17)14(11)20)3-10-9-6-8(18)1-2-13(9)19-15(10)21/h1-6,20H,(H,19,21)/b10-3-

InChI key

LMXYVLFTZRPNRV-KMKOMSMNSA-N

General description

A potent, cell-permeable, reversible, and ATP-competitive cRAF1 kinase inhibitor (IC50 = 9 nM). Shows ≥100-fold selectivity for Raf kinase versus Cdk1, Cdk2, c-Src, ERK2, MEK, p38, Tie2, VEGFR2, and c-Fms. Predicted broad spectrum antitumor agents. A 10 mM (500 µg/96 µl) solution of Raf1 Inhibitor I (Cat. No. 553003) in DMSO is also available.
A potent, cell-permeable, reversible, and ATP-competitive cRaf1 kinase inhibitor (IC50 = 9 nM). Shows ≥100-fold selectivity for Raf kinase versus Cdk1, Cdk2, c-src, ERK2, MEK, p38, Tie2, VEGFR2 and c-fms. cRaf1 inhibitors are predicted to be broad spectrum antitumor agents.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CRAFI kinase
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.







From Catalog:

Desc. Field- added "Predicted broad spectrum antitumor agents."

Other Notes

Lackey, K., et al. 2000. Bioorg. Med. Chem. Lett.10, 223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service