Skip to Content
MilliporeSigma

548000

1-Pyrrolidinecarbodithioic Acid, Ammonium Salt

Cell permeable. Inhibits the induction of NOS activity in rat alveolar macrophages. Prevents apoptosis in human HL-60 cells and thymocytes, but induces apoptosis in human and rat smooth muscle cells.

Synonym(s):

1-Pyrrolidinecarbodithioic Acid, Ammonium Salt, APDC, Ammonium Pyrrolidinedithiocarbamate, PDTC, NH4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H12N2S2
CAS Number:
Molecular Weight:
164.29
UNSPSC Code:
12352106
NACRES:
NA.77
MDL number:
Assay:
≥98% (titration)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S-]C(=S)N1CCCC1.[N+H4]

InChI

1S/C5H9NS2.H3N/c7-5(8)6-3-1-2-4-6;/h1-4H2,(H,7,8);1H3

InChI key

MDDIUTVUBYEEEM-UHFFFAOYSA-N

General description

Cell permeable. Inhibits the induction of nitric oxide synthase activity in rat alveolar macrophages. Prevents apoptosis in human HL-60 cells and thymocytes, but induces apoptosis in human and rat smooth muscle cells. Inhibits NF-κB activation and HIV replication in lymphocytes.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Induction of nitric oxide synthase activity in rat alveolar macrophages
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 3 months at -20°C.







From Catalog:

Desc. Field- added "cell-permeable"

Other Notes

Tsai, J.C., et al. 1996. J. Biol. Chem.271, 3667.
Pan, Z., et al. 1995. J. Biol. Chem.270, 7787.
Bessho, R., et al. 1994. Biochem. Pharmacol.48, 1883.
Sherman, M.P., et al. 1993. Biochem. Biophys. Res. Commun.191, 1301.
Schreck, R., et al. 1992. J. Exp. Med.175, 1181.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library