Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539648

Sigma-Aldrich

Staurosporine, N-Benzoyl

A cell-permeable Staurosporine derivative that displays antitumor properties.

Synonym(s):

Staurosporine, N-Benzoyl, PKG Inhibitor IX, N-Benzoylstaurosporine, CGP 41 251, PDGFR Tyrosine Kinase Inhibitor X, PKC412, PKG Inhibitor IX, N-Benzoylstaurosporine, CGP 41 251, PKC412, PDGFR Tyrosine Kinase Inhibitor X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H30N4O4
CAS Number:
Molecular Weight:
570.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 6 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]21c3c4[n](c7c(c4c8c(c3c9c2cccc9)CNC8=O)cccc7)[C@@H]5O[C@]1([C@@]([C@@H]5N(C)C(=O)c6ccccc6)(OC)C)C

InChI

1S/C35H30N4O4/c1-34(42-4)30(37(3)32(41)19-12-6-5-7-13-19)33-38-23-16-10-8-14-20(23)26-27-22(18-36-31(27)40)25-21-15-9-11-17-24(21)39(29(25)28(26)38)35(34,2)43-33/h5-17,30,33H,18H2,1-4H3,(H,36,40)/t30-,33-,34-,35+/m1/s1

InChI key

IIQIEMHSDLLZQA-QZPVEUDVSA-N

General description

A cell-permeable Staurosporine (Cat. No. 569397) derivative that displays antitumor properties. Acts as a broad-spectrum, reversible, and ATP-competitive inhibitor of PKC (α, β and γ), PDGFRβ, VEGFR2, Syk, PKCη, PKCδ, Flk-1, Flt3, Cdk1/B, PKA, c-Kit, c-Fgr, c-Src, VEGFR1, and EGFR (IC50 = 22 nM, 50 nM, 86 nM, 95 nM, 160 nM, 330 nM, 390 nM, 528 nM, 570 nM, 570 nM, 600 nM, 790 nM, 800 nM, 912 nM, and 1.0 µM, respectively). At higher concentrations, also inhibits Ins-R and IGF-1R (IC50 = 10 &microM). Shown to potently inhibit the proliferation of Ba/F3 cells expressing constitutively active Flt3 (IC50<10 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC (α, β and γ), PDGFRβ, VEGFR2, Syk, PKCη, PKCδ, Flk-1, Flt3, Cdk1/B, PKA, c-Kit, c-Fgr, c-Src, VEGFR1, and EGFR
Product competes with ATP.
Reversible: yes
Target IC50: 22 nM, 50 nM, 86 nM, 95 nM, 160 nM, 330 nM, 390 nM, 528 nM, 570 nM, 570 nM, 600 nM, 790 nM, 800 nM, 912 nM, and 1.0 µM against primary targets in the order listed

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Weisberg, E., et al. 2002. Cancer Cell1, 433.
Fabbro, D., et al. 2000. Anticancer Drug Des.15, 17.
Meyer, T., et al. 1999. Int. J. Cancer81, 669.
Meyer, T., et al. 1989. Int. J. Cancer43, 851.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bei Wang et al.
Redox biology, 65, 102825-102825 (2023-08-03)
Chemotherapeutic agents, such as doxorubicin (DOX), may cause cardiomyopathy, even life-threatening arrhythmias in cancer patients. Ferroptosis-an iron-dependent oxidative form of programmed necrosis, plays a pivotal role in DOX-induced cardiomyopathy (DIC). Prostaglandins (PGs) are bioactive signaling molecules that profoundly modulate cardiac
Oxana V Denisova et al.
Scientific reports, 12(1), 13796-13796 (2022-08-14)
Therapeutic resistance to kinase inhibitors constitutes a major unresolved clinical challenge in cancer and especially in glioblastoma. Multi-kinase inhibitors may be used for simultaneous targeting of multiple target kinases and thereby potentially overcome kinase inhibitor resistance. However, in most cases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service