Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C100H159N27O28
Molecular Weight:
2187.50
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
assay
≥95% (HPLC)
Quality Segment
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic), protect from light
color
off-white
solubility
DMSO: 5 mg/mL, water: 5 mg/mL
storage temp.
−20°C
General description
A cell-permeable hydrocarbon-stapled, α-helical peptide that mimics the native son of sevenless 1 (SOS1) helical domain (amino acids 929-944) with additional Arg-Arg tag for better cell permeability. Binds to wild-type and mutant KRAS at the SOS1 binding pocket in a sequence-specific manner and disrupts SOS1/KRAS interaction (EC50 = 79, 61, 103, 129, 140, and 124 nM for wild-type, G12D, G12V, G12C, G12S, and Q61H mutants, respectively) and blocks their nucleotide association. Reported to engage both GDP-KRAS and GTP-KRAS. Shown to impair the viability of cancer cells bearing G12D, G12C, G12V, G12S, G13D, and Q61H mutations (IC50 = 5 - 15 µM). Also diminishes the viability of HeLa and Colo320-HSR cells expressing wild-type KRAS. Inhibits the activity of MEK1/2, ERK1/2, and Akt in Panc10.05 cells treated with epidermal growth factor. Also shown to completely suppress the phosphorylation of Akt and ERK1/2 in Drosophila melanogaster expressing V12 mutant form of RAS85D upon RU486 (Cat. No. 475838) treatment.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
K-Ras
K-Ras
Reversible: yes
Packaging
Packaged under inert gas
Physical form
Supplied as a trifluoroacetate salt.
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Ac-RRFFGIXLTNXLKTEEGN (X=(S)-2-(4′-pentenyl)Ala, stapled)
Leshchiner, E. S. et al. 2015. Proc. Natl. Acad. Sci. USA.112, 1761.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.