Skip to Content
MilliporeSigma

5370

L-Proline

Synonym(s):

L-Proline, Pro

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
UNSPSC Code:
12352209
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - TW3584000
Merck USA index - 14, 7780

Quality Level

assay

≥98% (TLC)

form

powder, solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[N+H2]1C(CCC1)C(=O)[O-]

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)

InChI key

ONIBWKKTOPOVIA-UHFFFAOYSA-N

General description

A non-essential amino acid. Precursor of hydroxyproline in collagen.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under L-Proline


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
5370-100GM07790788051242