Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

5.33718

Sigma-Aldrich

NOD2 Signaling Inhibitor II, GSK717

Synonym(s):

NOD2 Signaling Inhibitor II, GSK717, N-(2-(1-(2-(2,3-Dihydro-1H-inden-5-ylamino)-2-oxoethyl)-1H-benzo[d]imidazol-2-yl)ethyl)-N-methylbenzamide, Nucleotide-binding Oligomerization Domain 2 Signaling Inhibitor II, MDP/NOD2-Mediated Signaling Inhibitor, GSK 717

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28N4O2
CAS Number:
Molecular Weight:
452.55
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable benzimidazole diamide compound that acts as a potent, selective, reversible and muramyl dipeptide (MDP)-induced NOD2 (MDP/nucleotide-binding oligomerization domain 2)-mediated signaling inhibitor (IC50 = 400 nM for MDP-stimulated IL-8 secretion in HEK293/hNOD2 cells). Does not affect NOD1, TNFR1, or TLR-2-mediated responses. Also shown to diminish synergy between NOD2 and TLR2. Reduces MDP-stimulated IL-8, IL-6, IL-1β, and TNFα release in primary human monocytes (~5 µM), but does not affect Pam2CSK4-stimulated IL-8 release. Due to a lack of typical activation loop/hinge-binding motif it is unlikely to act as a kinase inhibitor. NOD1 Inhibitor, Nodinitib-1 (>Cat. No. 488004) is also available.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A cell-permeable benzimidazole diamide compound that selectively inhibits muramyl dipeptide/MDP-induced, nucleotide-binding oligomerization domain 2/NOD2-mediated cytokine production (% inhibiton/cytokine = 91%/IL-8, 89%/IL-6, 87%/IL-1β, 80%/TNFα 25 h post 5 µM drug addition & 24 h post 100 ng/mL MDP stimulation) in primary human monocyte cultures, but not but not TLR2/6 agonist Pam2CSK4- (100 pg/mL) stimulated IL-8 secretion even at concentrations as high as 16 µM. Based on selectivity studies using the less potent, non-N-methylated analog GSK669, GSK717 is not expected to act as a kinase inhibitor or display significant potency against MDP-independent NOD2 activation, TNFα receptor- and NOD1-mediated cytokine productions. A great complement to the NOD1 signaling-selective inhibitor Nodinitib-1 (Cat. No. Cat. No. 488004). The exact mechanism of inhibition is not yet determined.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NOD2
Reversible: yes
Target IC50: 400 nM for MDP-stimulated IL-8 secretion in HEK293/hNOD2 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Rickard, D.J., et al. 2013. PLoS ONE.D408, e69619.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service