Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33259

Sigma-Aldrich

FOXM1 Inhibitor III, FDI-6

Synonym(s):

FOXM1 Inhibitor III, FDI-6, FDI6, Forkhead Domain Inhibitory Compound 6, NCGC00099374

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H11F4N3OS2
CAS Number:
Molecular Weight:
437.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c2c([s]c(c2N)C(=O)Nc4ccc(cc4)F)nc(c1)c3[s]ccc3

InChI

1S/C19H11F4N3OS2/c20-9-3-5-10(6-4-9)25-17(27)16-15(24)14-11(19(21,22)23)8-12(26-18(14)29-16)13-2-1-7-28-13/h1-8H,24H2,(H,25,27)

InChI key

ZATJMMZPGVDUOM-UHFFFAOYSA-N

General description

A cell-permeable thienopyridinylcarboxamide compound that targets forkhead box M1 (FOXM1) winged helix DNA-binding domain (DBD; aa 222-360 in FOXM1c) in a 1:1 stoichiometric ratio and effectively blocking FOXM1 DBD DNA-binding activity (IC50 = 22.5 µM by EMSA; 15 nM DNA probe & 750 nM FOXM1 DBD) and displays antiproliferative activity in breast cancer cultures (GI50 = 18 µM/MCF-7, 18.1 µM/PEO-1, and 21.8 µM/MDA-MB-231 with 72 h treatment). FDI-6 culture treatment reduces FOXM1 target genes promoter occupancy (by 36%/CDC25B and 38%/CCNB1 with 6 h treatment at 40 µM in MCF-7 cultures) and overall FOXM1 chromatin association (by 43%/20 µM and 79%/40 µM in MCF-7 cultures), resulting in decreased FOXM1 target genes mRNA level (by 57%/PLK1, 55%/CDC25B, 52%/AURKB, 45%/CARM1 with 6 h treatment at 40 µM in MCF-7 cultures). Close to a 19-fold enriched FOXM1 targets is seen among downregulated mRNA species when compared to those not affected by FDI-6 treatment, while little or no enrichment is seen pertaining to targets of highly homologous FOX family transcription factors FOXA1, FOXA2, and FOXP2 among mRNAs affected by FCI-6 treatment. Unlike Thiostrepton (Cat. No. 598226), FDI-6 does not inhibit 20S proteasome activity.
FOXM1 inhibitor FDI-6; NCGC00099374

Biochem/physiol Actions

Cell permeable: yes
Primary Target
FOXM1

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gormally , M.V., et al. 2014. Nat. Commun.5, 5156.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service