Skip to Content
MilliporeSigma

5.32606

5-Lipoxygenase Inhibitor, MK591

Synonym(s):

5-Lipoxygenase Inhibitor, MK591, FLAP Inhibitor, MK591, sodium 3-(3-(tert-butylthio)-1-(4-chlorobenzyl)-5-(quinolin-2-ylmethoxy)-1H-indol-2-yl]-2,2-dimethylpropanoate, L-686,708, MK-0591

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H34ClN2NaO3S
CAS Number:
Molecular Weight:
609.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

Quality Segment

form

solid

potency

3.1 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

SMILES string

[Na+].S(C(C)(C)C)c1c2c([n](c1CC(C)(C)C(=O)[O-])Cc5ccc(cc5)Cl)ccc(c2)OCc3nc4c(cc3)cccc4

InChI

1S/C34H35ClN2O3S.Na/c1-33(2,3)41-31-27-18-26(40-21-25-15-12-23-8-6-7-9-28(23)36-25)16-17-29(27)37(20-22-10-13-24(35)14-11-22)30(31)19-34(4,5)32(38)39;/h6-18H,19-21H2,1-5H3,(H,38,39);/q;+1/p-1

InChI key

YPURUCMVRRNPHJ-UHFFFAOYSA-M

General description

A cell permeable, bioavailable, 2-indolealkanoic acid derived compound that acts as a highly potent and selective inhibitor of leukotriene biosynthesis (IC50 = 3.1 nM in intact human polymorphonuclear leukocytes). Shown to inhibit 5-lipoxygenase (5-LOX) activity in LNCaP prostate cancer cells by high-affinity binding to 5-lipoxygenase activating protein (FLAP; IC50 = 1.6 nM in a FLAP binding assay). Induces apoptosis in LNCaP cells without blocking PI 3 kinase-Akt or ERK activities. Also shown to induce rapid phosphorylation of c-Jun N-terminal Kinase (JNK) and indirectly block PKCε activity. Suppresses the deposition of Ab1-40 and Ab1-42 peptides in Tg2576 mice (~40 mg/kg/day in diet) with concomitant reduction in the levels of presenilin 1, nicastrin, APH-1 and Pen-2, but without affecting notch signaling. Exhibits desirable pharmacokinetic properties with tmax of 3.3 h and t1/2 = ~12 h.
MK591; MK0591; 5-LOX inhibitor, 5- Lipoxygenase inhibitor, FLAP

Biochem/physiol Actions

Cell permeable: yes
Primary Target
FLAP

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chu, J., et al. 2012. J. Neuroinflamm.9, 127.
Sarveswaran, S., et al. 2011. Bioch. Biophy. Acta.1813, 2108.

Sarveswaran, S., et al. 2010. Cancer Lett.291, 167.
Uematsu, T., et al. 1995. Br. J. Clin. Pharm.40, 59.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library