Skip to Content

5.31996

KDM4A/KDM4B Inhibitor, NSC636819

Synonym(s):

KDM4A/KDM4B Inhibitor, NSC636819, JMJD2 Inhibitor, Jumonji C-containing Histone Lysine Demethylase Inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H12Cl4N2O4
CAS Number:
Molecular Weight:
510.15
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

Quality Segment

form

solid

potency

3.0 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

brown

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell permeable dinitrobenzene derivative that acts as a competitive inhibitor of KDM4A (IC50 = 6.4 µM; Ki = 5.5 µM) and KDM4B (IC50 = 9.3 µM; Ki = 3.0 µM) and completely blocks their demethylating activity toward H3K9me3 (~ 5 µM). Exhibits much reduced activities against KDM4D and KDM4E. Effectively induces apoptosis in LNCaP cells (IC50 = 16.5 µM after 3-day culture), but does not affect normal PNT2 cells (~ 5 to 20 µM over 6 days). Shown to up-regulate RB1 and CDH1 tumor suppressor genes and down-regulates IGF1R, FGFR3, CCNE2, AURKA, and AURKB oncogenes.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
KDM4A/B
Reversible: yes

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chu, C.H., et al. 2014. Journ. Med. Chem.57, 5975.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library