Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.31372

Sigma-Aldrich

Caspase-1/4 Inhibitor, VX-765

Synonym(s):

Caspase-1/4 Inhibitor, VX-765, ( S)-1-(( S)-2-((1-(4-Amino-3-chloro-phenyl)-methanoyl)-amino)-3,3-dimethyl-butanoyl)-pyrrolidine-2-carboxylic acid ((2 R,3 S)-2-ethoxy-5-oxo-tetrahydro-furan-3-yl)-amide, ( S)-1-(( S)-2-(4-Amino-3-chlorobenzamid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33ClN4O6
CAS Number:
Molecular Weight:
509.00
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

SMILES string

Clc1c(ccc(c1)C(=O)N[C@@H](C(C)(C)C)C(=O)N2[C@@H](CCC2)C(=O)N[C@@H]3[C@@H](OC(=O)C3)OCC)N

InChI

1S/C24H33ClN4O6/c1-5-34-23-16(12-18(30)35-23)27-21(32)17-7-6-10-29(17)22(33)19(24(2,3)4)28-20(31)13-8-9-15(26)14(25)11-13/h8-9,11,16-17,19,23H,5-7,10,12,26H2,1-4H3,(H,27,32)(H,28,31)/t16-,17-,19+,23+/m0/s1

InChI key

SJDDOCKBXFJEJB-MOKWFATOSA-N

General description

An enhanced cell-permeable, non-toxic, orally available masked aspartaldehyde prodrug that is converted to its active metabolite, VRT-043198, both in vitro and in vivo via hydrolytic cleavage by esterases. The active metabolite displays anti- inflammatory and anticonvulsant properties. Binds to the active site of caspase-1 and caspase-4 with high affinity to inhibit their activity (Ki = 800 pM and 600 pM, respectively). Does not affect the activities of trypsin, cathepsin B and has much reduced effect on other caspases (Ki = 100 nM to 21.5 µM) and granzyme B (Ki = 900 nM). Recently shown to inhibit pyroptosis and prevent CD4 T-cell death by HIV-1. Also shown to block the release of lipopolysaccharide- induced IL-1β and IL-18 in human PBMC (IC50 = 700 pM). Blocks Staphalococcus aureus - Cowen strain I-stimulated production of IL-1β, IL-18, and IFN-γ in human PBMCs (IC50 = 870 nM and 2.8 and 5.6 µM, respectively), but does not affect the TNF-α levels (IC50 >50 µM). Appears to cross the blood brain barrier to block seizure-induced IL-1β production and delay the onset and reduces the duration of seizures in rats (50 mg/kg, i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
The cell-permeable prodrug of the caspase-1/4-selective inhibitor VRT-43198 (Ki in nM = <0.6/Caspase-4, 0.8/Caspase-1, 100/Caspase-8, 560/Caspase-6, 1,030/Caspase-9, 9,000/Granzyme B, 16,000/Caspase-7, 21,500/Caspase-3; IC50 >100 µM against Cathepsin B & Trypsin). In addition to inhibiting LPS-induced IL-1β production in primary human PBMC cultures (IC50 ~1 µM), VX-765 is also effective in preventing HIV infection-induced IL-1β production and pyroptosis of CD4 T cells in human lymphoid aggregate cultures (HLAC; CD4 population = 29.2% in non-infected control cultures, 8.3% vs. 30.2% in infected cultures with or without 5 µM VX-765). VX-765 is orally available in mice (Blood Cmax = 0.78 µg/mL = 1.53 µM; Tmax = 1.0 h; AUClast = 2.06 µg · h/mL; 84 mg/kg, p.o.) and shown to display in vivo anti-inflammatory efficacy against LPS-induced plasma IL-1β production (EDmax = 100 mg/kg, p.o.), Oxazolone-induced delayed-type hypersensitivity (EDmax = 50 mg/kg, p.o.), collagen-induced arthritis (EDmax = 100 mg/kg, p.o.). When administered via intraperitoneal injection, VX-765 is also demonstrated to suppress the severity of seizure induction (EDmax = 50 mg/kg i.p.) among rats receiving kainic acid via intracerebroventricular injection.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-1/4
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Doitsh, G., et al. 2014. Nature505, 509.
Wannamaker, W., et al. 2007. J. Pharmacol. Exp. Ther.321, 509.

Ravizza, T., et al. 2006. Epilepsia47, 1160.
Stack, J.H., et al. 2005. J. Immunol.175, 2630.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service