Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.30151

Sigma-Aldrich

ATGL Inhibitor, Atglistatin

Synonym(s):

ATGL Inhibitor, Atglistatin, 3-(4ʹ-(Dimethylamino)biphenyl-3-yl)-1,1-dimethylurea, Adipose Triglyceride Lipase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3O
CAS Number:
Molecular Weight:
283.37
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

potency

700 nM IC50
355 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable biphenyldimethyl urea compound that inhibits adipose triglyceride lipase (ATGL) activity (IC50 = 700 nM; [Substrate] = [Triolein] = 165 µM) in a substrate-competitive (Ki = 355 nM) and highly selective manner, exhibiting little potency against hormone-sensitive lipase (HSL), monoglyceride lipase (MGL), pancreatic lipase, lipoprotein lipase, or lysophospholipases PNPLA6/7 even at concentrations as high as 100 µM. Shown to effectively suppress FA/glycerol productions from adipocytes (ICmax = 50 to 100 µM in 3T3-L1-differentiated adipocytes or primary murine white adipose tissue/WAT cultures) in vitro and in blood of mice in vivo via either oral or intraperitoneal administration (EDmax = 200 µmol/kg) without affecting plasma glucose, total cholesterol, insulin, or β-hydroxybutyrate (beta-HB) level. Significant cytotoxicity is reported at concentrations >50 µM in AML-12 murine hepatocyte cultures. Although ATGL deficiency causes cardiomyopathy in human and ATGL-ko animals due to severe triacylglycerol (TG) accumulation in cardiac muscle, significant atglistatin level (>0.2 nmol/g tissue) is only seen in liver, white and brown adipose tissues, and detectable TG accumulation is only observed in liver tissue in wt mice 8 h post single 200 µmol/kg oral dosage.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ATGL
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Mayer, N., et al. 2013. Nat. Chem. Biol.9, 785.
Produced under the license of University of Graz/Graz University of Technology

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service