Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30030

Sigma-Aldrich

Glutaminase Inhibitor II, BPTES

Synonym(s):

Glutaminase Inhibitor II, BPTES, N,Nʹ-(5,5ʹ-(2,2ʹ-Thio bis(ethane-2,1-diyl)) bis(1,3,4-thiadiazole-5,2-diyl)) bis(2-phenylacetamide), Kidney-Type Glutaminase Inhibitor II, GAC Inhibitor II, GLS1 Inhibitor II, KGA Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24N6O2S3
CAS Number:
Molecular Weight:
524.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥94% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

[s]1c(nnc1CCSCCc3[s]c(nn3)NC(=O)Cc4ccccc4)NC(=O)Cc2ccccc2

InChI

1S/C24H24N6O2S3/c31-19(15-17-7-3-1-4-8-17)25-23-29-27-21(34-23)11-13-33-14-12-22-28-30-24(35-22)26-20(32)16-18-9-5-2-6-10-18/h1-10H,11-16H2,(H,25,29,31)(H,26,30,32)

InChI key

MDJIPXYRSZHCFS-UHFFFAOYSA-N

General description

Glutaminase Inhibitor II,Bis-2-(5-phenylacetamido-1,2,4-thiadiazol-2-yl)ethyl sulfide (BPTES)is a cell-permeable bis-thiadiazole compound that selectivelyinhibits glutaminase-1(GLS1). It does not inhibit the liver-type GLS-2 mitochondrialglutaminase activity. It exerts its effect by inducing an inactive GLS1tetrameric conformation via a 2:1 inhibitor-to-tetramer stoichiometric binding.BPTES blocks the growth under aerobic/ normoxic conditions, and promotes celldeath under hypoxic conditions in P493 B-cell lymphoma cultures invitro, and effectively suppresses P493 tumor expansion in mice invivo.BPTES has been observedto be active in several cancer cells as it prevents cancer cellproliferation.

Application

Glutaminase Inhibitor II, BPTES has been used asa glutaminase inhibitor to study its effects on the release of extracellularvesicles (EVs) in human immunodeficiency virus (HIV-1) infected macrophages.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
kidney-type glutaminase
Reversible: yes
Target IC50: 140 and 210 nM in HEK293 cells expressing wt-KGA and F318Y-cKGA, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Shukla, K., et al. 2012. J. Med. Chem.55, 10551.

Thangavelu, K., et al. 2012. Proc. Natl. Acad. Sci. USA109, 7705.
Le, A., et al. 2012. Cell Metab.15, 110.
DeLaBarre, B., et al. 2011. Biochemistry 50, 10764.
Seltzer, M.J., et al. 2010. Cancer Res.70, 8981.
Robinson, M.M., et al. 2007. Biochem. J.406, 407.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glutaminase 1 regulates the release of extracellular vesicles during neuroinflammation through key metabolic intermediate alpha-ketoglutarate
Beiqing W, et al.
Journal of Neuroinflammation (2018)
Charles J McDonald et al.
Neurochemistry international, 88, 10-14 (2014-12-17)
The GLS1 gene encodes a mitochondrial glutaminase that is highly expressed in brain, kidney, small intestine and many transformed cells. Recent studies have identified multiple lysine residues in glutaminase that are sites of N-acetylation. Interestingly, these sites are located within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service