Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528822

Sigma-Aldrich

PARP Inhibitor XII

The PARP Inhibitor XII, also referenced under CAS 489457-67-2, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

PARP Inhibitor XII, 3-(4-Chlorophenyl)quinoxaline-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClN3O
CAS Number:
Molecular Weight:
283.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light brown

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)c2nc3c(nc2)cccc3C(=O)N

InChI

1S/C15H10ClN3O/c16-10-6-4-9(5-7-10)13-8-18-12-3-1-2-11(15(17)20)14(12)19-13/h1-8H,(H2,17,20)

InChI key

FLYGLPYJEQPCFY-UHFFFAOYSA-N

General description

A potent quinoxaline-based PARP inhibitor with a ~5-fold selectivity towards PARP-2 over PARP-1 (IC50 = 7 and 33 nM, respectively). Reported to be brain-permeant and exhibit good pharmacokinetics.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Ishida, J., et al. 2006. Bioorg. Med. Chem.14, 1378.
Iwashita, A., et al. 2005. FEBS Lett.579, 1389.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service