Skip to Content
MilliporeSigma

528283

Polo-like Kinase Inhibitor II, BTO-1

The Polo-like Kinase Inhibitor II, BTO-1, also referenced under CAS 40647-02-7, controls the biological activity of Polo-like Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Polo-like Kinase Inhibitor II, BTO-1, 5-Cyano-7-nitro-2-(benzothiazolo-N-oxide)-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H4N4O4S
CAS Number:
Molecular Weight:
264.22
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

beige to brown

solubility

DMSO: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c2c([n+](c1C(=O)N)[O-])cc(cc2[N+](=O)[O-])C#N

InChI

1S/C9H4N4O4S/c10-3-4-1-5-7(6(2-4)13(16)17)18-9(8(11)14)12(5)15/h1-2H,(H2,11,14)

InChI key

FKRBAXZAJBBIAJ-UHFFFAOYSA-N

General description

A cell-permeable benzothiazolo-N-oxide compound that targets the ATP-binding pocket of polo-like kinase and is shown to inhibit Plk1 kinase activity in cell-free kinase assays (IC50 = 8.0 µM) and suppress the phosphorylation of cellular Plk1 substrate Cdc25C in rat kangaroo kidney-derived PTK cells (75% inhibition at 6.3 µM). BTO-1 treatment of cultured cells results in mitosis defects consistent with loss-of-Plk1 phenotypes seen in Plk1 RNAi-treated U20S cells, including the appearance of monopolar spindles and the reduction of γ-tubulin at the centrosomes. Plk1 inhibition by BTO-1 in HeLa cells results in a blockage of Rho and Rho-GEF recruitment, which is essential for the assembly of a functional contractile ring.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Brennan, I.M., et al. 2007. PloS ONE2, e409.
Peters, U., et al. 2006. Nat. Chem. Biol.2, 618.
McInnes, C., et al. 2006. Nat. Chem. Biol.2, 608.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library