Skip to Content
MilliporeSigma
All Photos(1)

Documents

527948

Sigma-Aldrich

Piceatannol

A cell-permeable, substrate competitive and reversible plant metabolite that inhibits the activity of rat liver protein kinase A catalytic subunit (IC₅₀ = 3 µM), PKC (IC₅₀ = 8 µM), MLCK (IC₅₀ = 12 µM).

Synonym(s):

Piceatannol, trans-3,3ʹ,4,5ʹ-Tetrahydroxystilbene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O4
CAS Number:
Molecular Weight:
244.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H12O4/c15-11-5-10(6-12(16)8-11)2-1-9-3-4-13(17)14(18)7-9/h1-8,15-18H/b2-1+

InChI key

CDRPUGZCRXZLFL-OWOJBTEDSA-N

General description

A cell-permeable, substrate competitive and reversible plant metabolite that inhibits the activity of rat liver protein kinase A catalytic subunit (IC50 = 3 µM), PKC (IC50 = 8 µM), MLCK (IC50 = 12 µM). In RBL-2H3 cells, the selective inhibition of Syk by piceatannol results in inhibition of FceR1-mediated signaling. Also inhibits wheat embryo Ca2+-dependent protein kinase (CDPK) (IC50 = 19 µM), and p72syk (IC50 = 10 µM), a non-receptor tyrosine kinase, relative to Lyn in isolated enzyme preparations. Also possesses anti-leukemic properties. Reported to activate sirtuins and promote the survival of eukaryotic cells.
A cell-permeable, substrate competitive, and reversible plant metabolite that inhibits the activity of rat liver protein kinase A catalytic subunit (IC50 = 3 µM), PKC (IC50 = 8 µM), MLCK (IC50 = 12 µM). In RBL-2H3 cells, the selective inhibition of Syk by piceatannol results in inhibition of FceR1-mediated signaling. Also inhibits wheat embryo Ca2+-dependent protein kinase (CDPK) (IC50 = 19 µM), and p72syk (IC50 = 10 µM), a non-receptor tyrosine kinase, relative to Lyn in isolated enzyme preparations. Also possesses anti-leukemic properties. Reported to activate sirtuins and promote the survival of eukaryotic cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Activity of rat liver protein kinase A catalytic subunit
Product competes with ATP.
Reversible: yes
Target IC50: 3 µM, 8 µM, and 12 µM against the activity of rat liver protein kinase A catalytic subunit, PKC, and MLCK, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot, flush with an inert gas, and freeze (-20°C). Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Howitz, K.T., et al. 2003. Nature425, 191.
Wang, B.H., et al. 1998. Planta Med. 64, 195.
Keely, P.J., and Parise, L.V. 1996. J. Biol. Chem. 271, 26668.
Oliver, J.M., et al. 1994. J. Biol. Chem. 269, 29697.
Thakkar, K., et al. 1993. J. Med. Chem. 36, 2950.
Geahlen, R.L., et al. 1989. Biochem. Biophys. Res. Commun. 165, 241.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Nasi et al.
Annals of the rheumatic diseases, 75(7), 1372-1379 (2015-08-09)
Basic calcium phosphate (BCP) crystal and interleukin 6 (IL-6) have been implicated in osteoarthritis (OA). We hypothesise that these two factors may be linked in a reciprocal amplification loop which leads to OA. Primary murine chondrocytes and human cartilage explants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service