Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze
description
Merck USA index - 14, 7147
Quality Segment
assay
≥98% (HPLC)
form
solid
optical activity
[α]20.0/D -90.0±5.0, c = 0.5 in methanol
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
DMSO: 1 mg/mL, methanol: 1 mg/mL
storage temp.
2-8°C
SMILES string
N([C@@H](C(C)C)C(=O)NC(C(O)CC(=O)N[C@@H](C)C(=O)NC(C(O)CC(=O)O)CC(C)C)CC(C)C)C(=O)[C@@H](NC(=O)CC(C)C)C(C)C
InChI
1S/C34H63N5O9/c1-17(2)12-23(37-33(47)31(21(9)10)39-34(48)30(20(7)8)38-27(42)14-19(5)6)25(40)15-28(43)35-22(11)32(46)36-24(13-18(3)4)26(41)16-29(44)45/h17-26,30-31,40-41H,12-16H2,1-11H3,(H,35,43)(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,44,45)/t22-,23?,24?,25?,26?,30-,31-/m0/s1
InChI key
FAXGPCHRFPCXOO-JHTFMCFMSA-N
General description
A reversible inhibitor of aspartic proteases. Inhibits cathepsin D, pepsin, and renin. Reported to block apoptosis induced by CA-074 (Cat. No. 508437).
Effective concentration: 1.0 µM.
Biochem/physiol Actions
EC50 = 1.0 µM in blocking apoptosis induced by CA-074 in PC12 cell lines
Primary Target
cathepsin D
cathepsin D
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.
Other Notes
Isahara, K., et al. 1999. Neuroscience91, 233.
Kanamori, S., et al. 1998. Biochem. Biophys. Res. Commun.251, 204.
Szewczuk, Z., et al. 1992. Int. J. Pept. Prot. Res.40, 233.
Carraway, R.E., et al. 1991. Peptides12, 107.
Eid, M., et al. 1981. Biochem. J.197, 465.
Umezawa, H., et al. 1970. J. Antibiot.23, 259.
Kanamori, S., et al. 1998. Biochem. Biophys. Res. Commun.251, 204.
Szewczuk, Z., et al. 1992. Int. J. Pept. Prot. Res.40, 233.
Carraway, R.E., et al. 1991. Peptides12, 107.
Eid, M., et al. 1981. Biochem. J.197, 465.
Umezawa, H., et al. 1970. J. Antibiot.23, 259.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.