Skip to Content
MilliporeSigma

512585

Parathyroid Hormone 1-34, Rat

Synonym(s):

Parathyroid Hormone 1-34, Rat, Parathyroid Hormone 1-34, Rat (rPTH 1-34), rPTH 1-34

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C180H291N55O48S2
CAS Number:
Molecular Weight:
4057.71
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 7034

Quality Segment

assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

5% acetic acid: 1 mg/mL, DMSO: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Synthetic, rat parathryroid hormone. Suitable for studies of bone metabolism and glandular disorders, characterization of specific receptor sites, and as an antigen.

Other Notes

Coltrera, M.D., et al. 1981. J. Biol. Chem.256, 10555.
Roos, B.A., et al. 1981. J. Clin. Endocrinol. Metab.53, 709.
Jubiz, W., et al. 1972. J. Clin. Invest.51, 2040.
H-Ala-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Ala-Ser-Val-Glu-Arg-Met-Gly-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OH

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library