Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09708

Sigma-Aldrich

SKF-97541

Synonym(s):

SKF-97541, GABA B Receptor Agonist, SKF-97541, SKF 97541, CGP 35024, 3-APMPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12NO2P
CAS Number:
Molecular Weight:
137.12
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 100 mM

storage temp.

2-8°C

SMILES string

[P](=O)(O)(CCC[N+H3])C

InChI

1S/C4H12NO2P/c1-8(6,7)4-2-3-5/h2-5H2,1H3,(H,6,7)/p+1

InChI key

NHVRIDDXGZPJTJ-UHFFFAOYSA-O

General description

A highly potent agonist of GABAB receptor. Depresses excitatory postsynaptic potentials mediated by glutamate in the striatum, and hyperpolarized neurones in the substantia nigra (IC50 = 92 nM). Shown to be about 10-times more potent than baclofen (Cat. No. 505873). Induces vasodepression in pulmonary vascular bed that is modulated by an ATP-sensitive K+ channel. Induces gastric acid secretion in a dose-dependent manner (0.01 - 3 mg/kg, i.v.) in mice.
A highly potent agonist of GABAB receptor. Depresses excitatory postsynaptic potentials mediated by glutamate in the striatum, and hyperpolarized neurones in the substantia nigra (IC50 = 92 nM). Shown to be about 10-times more potent than baclofen (Cat. No. 505873). Induces vasodepression in pulmonary vascular bed that is modulated by an ATP-sensitive K+ channel. Induces gastric acid secretion in a dose-dependent manner (0.01 - 3 mg/kg, i.v.) in mice.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
GABAB

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kaye, A.D., et al. 2004. Anesth. Analg.99, 758.
Piqueras, L., and Martinez, V. 2004. Br. J. Pharmacol.142, 1038.

Seabrook, G.R., et al. 1990. Br. J. Pharmacol.101, 949.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service