Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08957

Sigma-Aldrich

PDE5 Inhibitor, Cpd7a

Synonym(s):

PDE5 Inhibitor, Cpd7a

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20ClN3O2
CAS Number:
Molecular Weight:
393.87
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

270 pM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

cream

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, brain permeant, quinoline-based compound that acts as a highly potent and selective inhibitor of phosphodiesterase 5 (PDEV; IC50 = 270 pM). Exhibits over 10,000 fold greater selectivity over various other phosphodiestrases and trivial activity against PDE VI (IC50 = 339 nM). Shown to increase cGMP levels in mouse hippocampus and attenuates long term potentiation and memory defects in mice with elevated levels of oligomeric Aβ42. It is rapidly absorbed with peak plasma concentration observed at 30 min. Displays desirable pharmacokinetic properties with plasma t1/2 = 1.33 h and brain t1/2 = 1.04 h.
A cell-permeable, brain permeant, quinoline-based compound that acts as a highly potent and selective inhibitor of phosphodiesterase 5 (PDEV; IC50 = 270 pM). Exhibits over 10,000 fold greater selectivity over various other phosphodiestrases and trivial activity against PDE VI (IC50 = 339 nM). Shown to increase cGMP levels in mouse hippocampus and attenuates long term potentiation and memory defects in mice with elevated levels of oligomeric Aβ42. It is rapidly absorbed with peak plasma concentration observed at 30 min. Displays desirable pharmacokinetic properties with plasma t1/2 = 1.33 h and brain t1/2 = 1.04 h.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
PDE5
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fiorito, J., et al. 2013. Eur. J. Med. Chem.60, 285.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service