Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.08146

Sigma-Aldrich

WAY-100635 Maleate

Synonym(s):

WAY-100635 Maleate, 5-HT 1A Serotonis Receptor Antagonist, WAY-100635 Maleate, WAY 100635, WAY100635, 5-HT1A Serotonis Receptor Antagonist, WAY-100635 Maleate, WAY 100635, WAY100635

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34N4O2 · xC4H4O4
CAS Number:
Molecular Weight:
422.56 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 50 mM

storage temp.

2-8°C

InChI

1S/C25H34N4O2.C4H4O4/c1-31-23-12-6-5-11-22(23)28-18-15-27(16-19-28)17-20-29(24-13-7-8-14-26-24)25(30)21-9-3-2-4-10-21;5-3(6)1-2-4(7)8/h5-8,11-14,21H,2-4,9-10,15-20H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

XIGAHNVCEFUYOV-BTJKTKAUSA-N

General description

A selective 5-HT1A receptor antagonist (pKi = 7.9-9.2). WAY-100635 has been reported to have good inhibitory action centrally and peripherally (1mg/kg dose in rats) but has low intrinsic effects, making it a silent 5-HT1A receptor antagonist. WAY-100635 displaced specific binding of the 5-HT1A radioligand, [3H]8-OH-DPAT (8-hydroxy-2-(di-n-propylamino)tetralin), to rat hippocampal membranes with a pIC50 of 8.87. This represented a greater than 100-fold selectivity relative to binding at other 5-HT receptor subtypes and major neurotransmitter receptor, reuptake and ion channel sites. WAY-100635 has been used in a variety of vertebrate species in vivo and in vitro.

Biochem/physiol Actions

Primary Target
5-HT1A

Warning

Toxicity: Standard Handling (A)

Preparation Note

Warming is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hunt, G. E. et al. 2011. Brain Res. Bull.86, 65.

Caamano, R. I. et al. 2010. J. Exp. Biol.213, 1813.

Newman-Tancredi, A. et al. 1998. Naunyn Schmiedebergs Arch. Pharmacol.357, 205.
Forster, E. A. et al. 1995. Eur. J. Pharmacol.281, 81.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service