Skip to Content
MilliporeSigma
All Photos(1)

Documents

506600

Sigma-Aldrich

Pamidronate, Disodium Salt

A potent and specific inhibitor of bone resorption that does not cause any significant detrimental effect on bone growth and mineralization at concentrations used.

Synonym(s):

Pamidronate, Disodium Salt, 3-Amino-1-hydroxypropylidene-bisphosphonic Acid, 2Na, Pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9NO7P2 · 2Na · 5H2O
CAS Number:
Molecular Weight:
369.11
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C3H11NO7P2.2Na.5H2O/c4-2-1-3(5,12(6,7)8)13(9,10)11;;;;;;;/h5H,1-2,4H2,(H2,6,7,8)(H2,9,10,11);;;5*1H2/q;2*+1;;;;;/p-2

InChI key

CZYWHNTUXNGDGR-UHFFFAOYSA-L

General description

A calcium metabolism regulator. A potent and specific inhibitor of bone resorption at doses devoid of any significant detrimental effect on bone growth and mineralization. Effective in a variety of conditions characterized by pathologically enhanced bone turnover, including Paget′s disease, hypercalcemia of malignancy, osteolytic bone metastasis, and steroid-induced cell death in vitro. Provided in a hydrated form containing five molecules of water.
A potent and specific inhibitor of bone resorption that does not cause any significant detrimental effect on bone growth and mineralization at concentrations used. Effective in a variety of conditions characterized by pathologically enhanced bone turnover, including Paget’s disease, hypercalcaemia of malignancy, osteolytic bone metastasis, steroid-induced osteoporosis, and idiopathic osteoporosis. Inhibits breast cancer cell growth by inducing cell death in vitro. Provided in hydrated form, contains five molecules of water.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inhibitor of bone resorption
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Fromigue, O., et al. 2000. J. Bone Miner. Res.15, 2211.
Fitton, A., and McTravish, D. 1991. Drugs41, 289.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María A Günther Sillero et al.
FEBS letters, 580(24), 5723-5727 (2006-10-03)
T4 RNA ligase catalyzes the synthesis of ATP beta,gamma-bisphosphonate analogues, using the following substrates with the relative velocity rates indicated between brackets: methylenebisphosphonate (pCH(2)p) (100), clodronate (pCCl(2)p) (52), and etidronate (pC(OH)(CH(3))p) (4). The presence of pyrophosphatase about doubled the rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service