Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05991

Sigma-Aldrich

Flumazenil

Synonym(s):

Flumazenil, GABA A Receptor Antagonist, Flumazenil, Ro 15-1788, GABAA Receptor Antagonist, Flumazenil, Ro 15-1788

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14FN3O3
CAS Number:
Molecular Weight:
303.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mM

storage temp.

2-8°C

SMILES string

Fc1cc2c(cc1)[n]3c(c(nc3)C(=O)OCC)CN(C2=O)C

InChI

1S/C15H14FN3O3/c1-3-22-15(21)13-12-7-18(2)14(20)10-6-9(16)4-5-11(10)19(12)8-17-13/h4-6,8H,3,7H2,1-2H3

InChI key

OFBIFZUFASYYRE-UHFFFAOYSA-N

General description

A competitive blocker of benzodiazepine activation of inhibitory GABAergic synpases (ID50 = 200 µg/kg/i.p). Binds to the benzodiazepine site of GABAA receptors. Enhances GABAA-receptor mediated currents and antagonizes the enhancing effects of benzodiapine agonist flurazepam. Exhibits fast association and dissociation from the benzodiazepine binding site. Permeates the blood brain barrier, however, the level of uptake may depend on efflux effects of p-glycogprotein transporter.
A competitive blocker of benzodiazepine activation of inhibitory GABAergic synpases (ID50 = 200 mg/kg/i.p). Binds to the benzodiazepine site of GABAA receptors. Enhances GABAA-receptor mediated currents and antagonizes the enhancing effects of benzodiapine agonist flurazepam. Exhibits fast association and dissociation from the benzodiazepine binding site. Permeates the blood brain barrier, however, the level of uptake may depend on efflux effects of p-glycogprotein transporter.

Biochem/physiol Actions

Primary Target
GABAA

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Froklage, F.E., et al. 2012. EJNMMI Research.2, 12.
Li, J., et al. 2006. Neuropharmacol.51, 168.
Weiss, M., et al. 2002. Neurochem. Res.27, 1605.
Atack, J., et al. 1999. Neuropsychopharmacol.20, 255.
Votey, S.R., et al. 1991. Ann. Emerg. Med.20, 181.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service