Skip to Content
MilliporeSigma

Skip To

5.00952

all-rac-α-Tocopheryl acetate

(Vitamin E acetate), EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, FCC

Synonym(s):

all-rac-α-Tocopheryl acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
MDL number:
UNSPSC Code:
12352108
EC Index Number:
231-710-0
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, Ph. Eur., USP

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

liquid

autoignition temp.

303 °C

mp

-27.5 °C

transition temp

flash point 210 °C

density

0.96 g/cm3 at 20 °C

application(s)

pharmaceutical

storage temp.

15-25°C

SMILES string

O1[C@@](CCc2c1c(c(c(c2C)OC(=O)C)C)C)(CCC[C@@H](CCC[C@@H](CCCC(C)C)C)C)C

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

General description

As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

all-rac-α-Tocopheryl acetate (Vitamin E acetate) protects cells against free radicals. Vitamin E is used in skin and hair care products, as well as in dietary supplements. all-rac-α-Tocopheryl acetate (Vitamin E acetate) Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1030T16000001667701
description

(Vitamin E acetate), EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, FCC

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

European Pharmacopoeia (EP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

agency

BP, USP, Ph. Eur.

agency

traceable to Ph. Eur. T1600000, traceable to USP 1667701

agency

-

agency

USP

form

liquid

form

-

form

liquid

form

-

product line

EMPROVE® ESSENTIAL

product line

-

product line

-

product line

-

density

0.96 g/cm3 at 20 °C

density

0.96 g/mL at 20 °C (lit.)

density

0.96 g/mL at 20 °C (lit.)

density

0.96 g/mL at 20 °C (lit.)

reg. compliance

FCC

reg. compliance

-

reg. compliance

-

reg. compliance

-


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.