Skip to Content
MilliporeSigma

5.00667

SAFC

Thiamine phosphoric acid ester chloride dihydrate

EMPROVE® API

Pharma Manufacturing

Synonym(s):

Thiamine monophosphate chloride dihydrate, [Thiaminium dihydrogen phosphate chloride (estersalt)], Aneurine monophosphate chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18ClN4O4PS · 2H2O
CAS Number:
Molecular Weight:
416.82
Beilstein:
3844977
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-536-9

vapor pressure

<1 Pa ( 50 °C)

Quality Level

product line

EMPROVE® API

form

solid

pH

2-3 (20 °C, 100 g/L in H2O)

mp

135 °C (decomposition)

solubility

176 g/L

density

1.53 g/cm3 at 20 °C

bulk density

670 kg/m3

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-25°C

SMILES string

[Cl-].Cc1ncc(C[n+]2csc(CCOP(O)(O)=O)c2C)c(N)n1

InChI

1S/C12H17N4O4PS.ClH/c1-8-11(3-4-20-21(17,18)19)22-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H3-,13,14,15,17,18,19);1H

InChI key

GUGWNSHJDUEHNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Thiamine phosphoric acid ester chloride dihydrate is a source for Vitamin B1 for use in clinical nutrition, vitamin supplements and infusion for Vitamin B1 deficiency. Thiamine phosphoric acid ester chloride dihydrate Emprove® API is manufactured according to ICH Q7 guidelines. An ASMF is available.
This product is part of our proprietary Viscosity Reduction Platform and suitable for highly concentrated protein-based formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service