Skip to Content
MilliporeSigma

Skip To

494459

n-Octyl-β-D-glucopyranoside

CAS 29836-26-8 is a non-ionic detergent intended for solubilizing membrane-bound proteins in their native state and for the preparation of lipid vesicles.

Synonym(s):

n-Octyl-β-D-glucopyranoside, OG, n-Octylglucoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
UNSPSC Code:
12161902
NACRES:
NA.21
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

mol wt

292.38 g/mol

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

aggregation number

84

impurities

≤0.05% n-octanol, ≤35 mg/dL glucose

color

white

CMC

20 - 25 mM

mp

106.8 °C (at 984 hPa)

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OCCCCCCCC

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

HEGSGKPQLMEBJL-RKQHYHRCSA-N

General description

Absorbance (10%, H2O, 260 nm):≤0.3.
Non–ionic detergent intended for solubilizing membrane–bound proteins in their native state and for the preparation of lipid vesicles. Its well–defined chemical structure, small uniform micelles, and high water solubility make it superior to most other nonionic detergents for membrane solubilization. Because of its high critical micelle concentration (20-25 mM), it has the additional advantage of being removed easily by dialysis when compared to bile salts. Aggregation number: 84.
Nonionic detergent intended for solubilizing membrane-bound proteins in their native state and for the preparation of lipid vesicles. Its well-defined chemical structure, small uniform micelles, and high water solubility make it superior to most other nonionic detergents for membrane solubilization. Because of its high critical micelle concentration (20-25 mM), it has the additional advantage of being removed easily by dialysis when compared to bile salts. Aggregation number: 84.

Application


  • Tube-gel digestion: a novel proteomic approach for high throughput analysis of membrane proteins.: This article introduces a new method for analyzing membrane proteins using tube-gel digestion, which employs n-Octyl-β-D-glucopyranoside for effective solubilization of membrane proteins. This approach enhances the throughput and efficiency of proteomic analyses (Lu et al., 2005).

  • Mixed micellization of a nonionic-cationic surfactant system constituted by n-octyl-beta-D-glucopyranoside/dodecyltrimethylammonium bromide/H2O.: The study explores the micellization behavior of a nonionic-cationic surfactant system involving n-Octyl-β-D-glucopyranoside and its interactions. The findings are significant for understanding surfactant properties and applications in biochemical processes (del Burgo et al., 2004).

Preparation Note

Following reconstitution, store in the refrigerator (4°C), under sterile conditions. Aqueous stock solutions are stable for up to 3 months at 4°C. Solutions are susceptible to hydrolysis (especially acidic solutions) and enzymatic hydrolysis.

Other Notes

Hoshino, T., et al. 1992. J. Biol. Chem.267, 21313.
Levy, D., et al. 1992. Biochim. Biophys. Acta1107, 283.
Nunn, R.S., et al. 1992. J. Mol. Biol.228, 1259.
Lorber, B., et al. 1990. Biochim. Biophys. Acta1023, 254.
Jopski, B., et al. 1989. Biochim. Biophys. Acta978, 79.
Camm, E.L., et al. 1981. Plant Physiol.67, 1061.
Gould, R.J., et al. 1981. Biochemistry20, 6776.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
O988210634425001O8001
description

CAS 29836-26-8 is a non-ionic detergent intended for solubilizing membrane-bound proteins in their native state and for the preparation of lipid vesicles.

description

BioXtra, ≥98% (GC)

description

non-ionic

description

≥98% (GC)

assay

≥98% (HPLC)

assay

≥98% (GC)

assay

99% (GC)

assay

≥98% (GC)

form

solid

form

powder

form

-

form

powder

aggregation number

84

aggregation number

84

aggregation number

84

aggregation number

84

storage temp.

15-25°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

solubility

water: 100 mg/mL

solubility

-

solubility

-

solubility

-

mp

106.8 °C (at 984 hPa)

mp

-

mp

-

mp

-


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters