Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

487920

Sigma-Aldrich

S-Nitrosoglutathione

≥95% (HPLC), solid, nitric oxide carrier, Calbiochem®

Synonym(s):

S-Nitrosoglutathione, GSNO, SNOG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N4O7S
CAS Number:
Molecular Weight:
336.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

S-Nitrosoglutathione, A utility carrier of nitric oxide.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pink

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C10H16N4O7S/c11-5(10(19)20)1-2-7(15)13-6(4-22-14-21)9(18)12-3-8(16)17/h5-6H,1-4,11H2,(H,12,18)(H,13,15)(H,16,17)(H,19,20)/t5-,6-/m0/s1

InChI key

HYHSBSXUHZOYLX-WDSKDSINSA-N

General description

A utility carrier of nitric oxide. Breaks down at physiological pH to produce nitric oxide. Causes smooth muscle relaxation and inhibits platelet aggregation. Induces apoptosis in mouse thymocytes. Note: 1 set = 5 x 2.5 mg.
A utility carrier of nitric oxide. Breaks down to produce nitric oxide and a glutathione radical at pH 7.4. Causes smooth muscle relaxation and inhibits platelet aggregation. Induces apoptosis in mouse thymocytes. Note: 1 set = 5 x 2.5 mg.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A utility carrier of nitric oxide
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C or for up to 2 h at 4°C.
Reconstitute in cold distilled H₂O (4°C) free of divalent cations other than Ca2+.

Other Notes

Sandau, K., et al. 1996. Cell Signal.8, 173.
Askew, S.C. 1995. Bioorg. Med. Chem.3, 1.
Fehsel, K., et al. 1995. J. Immunol.155, 2858.
de Belder, A.J., et al. 1994. Cardiovasc. Res.28, 691.
Sexton, D.J., et al. 1994. Photochem. Photobiol.59, 463.
Singh, R.J., et al. 1994. FEBS Lett. 360, 47.
Merryman, P.F., et al. 1993. Arthritis Rheum.36, 1414.
Park, J.W., et al. 1993. Biochem. Mol. Biol. Int.30, 885.
Radomski, M.W., et al. 1992. Br. J. Pharmacol.107, 745.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service